Assay Details
Gene Symbol: MIR3955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107888.1 | 1 | 53 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-370 | MIR370 | oar-mir-370 | -84.7kb | Sense | Chr. 18 - 66332610 - 66332715 on NCBI Build Oar_rambouillet_v1.0 |
Downstream | microRNA mir-379 | MIR379 | oar-mir-379 | 21.2kb | Sense | Chr. 18 - 66438646 - 66438770 on NCBI Build Oar_rambouillet_v1.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
