Assay Details
Gene Symbol: MIR380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107892.1 | 1 | 49 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-299 | MIR299 | oar-mir-299 | -1.2kb | Sense | Chr. 18 - 66440407 - 66440460 on NCBI Build Oar_rambouillet_v1.0 |
Downstream | microRNA mir-411b | MIR411B | oar-mir-411b | 59.0 | Sense | Chr. 18 - 66441775 - 66441875 on NCBI Build Oar_rambouillet_v1.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
