Assay Details
Gene Symbol: MIR3956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_107911.1 | 1 | 59 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-1185 | MIR1185 | oar-mir-1185 | -629.0 | Sense | Chr. 18 - 66458215 - 66458320 on NCBI Build Oar_rambouillet_v1.0 |
Downstream | microRNA mir-381 | MIR381 | oar-mir-381 | 799.0 | Sense | Chr. 18 - 66459876 - 66460003 on NCBI Build Oar_rambouillet_v1.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
