Assay Details
Gene Symbol: RIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002708103.3 | XP_002708149.1 | 20 - 21 | 3265 | 72 | |
XM_008255095.2 | XP_008253317.1 | 20 - 21 | 3265 | 72 | ||
XM_008255104.2 | XP_008253326.1 | 20 - 21 | 3262 | 72 | ||
XM_017341498.1 | XP_017196987.1 | 20 - 21 | 3262 | 72 | ||
XM_017341502.1 | XP_017196991.1 | 19 - 20 | 3157 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
