Assay Details
Gene Symbol: ANAPC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008272209.2 | XP_008270431.1 | 10 - 11 | 1892 | 68 | |
XM_008272210.2 | XP_008270432.1 | 10 - 11 | 1844 | 68 | ||
XM_017349517.1 | XP_017205006.1 | 9 - 10 | 1296 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
