Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718897.3 | XP_002718943.3 | 5 - 6 | 1723 | 74 | |
XM_017348925.1 | XP_017204414.1 | 5 - 6 | 1723 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
