Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002713858.3 | XP_002713904.1 | 4 - 5 | 712 | 84 | |
XM_017344333.1 | XP_017199822.1 | 3 - 4 | 478 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
