Assay Details
Gene Symbol: KDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195670.1 | NP_001182599.1 | 25 - 26 | 68 | ||
XM_017347243.1 | XP_017202732.1 | 25 - 26 | 68 | |||
GenBank mRNA | AB017155.1 | - | 3 - 4 | 351 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
