Assay Details
Gene Symbol: LOC100008716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717113.3 | XP_002717159.1 | 3 - 4 | 494 | 85 | |
GenBank mRNA | AF123437.1 | - | 1 - 2 | 101 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
