Assay Details
Gene Symbol: VIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717420.3 | XP_002717466.1 | 3 - 4 | 876 | 86 | |
GenBank mRNA | AY465353.1 | - | 1 - 2 | 103 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
