Assay Details
Gene Symbol: LOC100009591 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712249.3 | XP_002712295.1 | 8 - 9 | 1595 | 93 | |
GenBank mRNA | DQ242485.1 | - | 2 - 3 | 196 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
