Assay Details
Gene Symbol: PROC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195801.1 | NP_001182730.1 | 7 - 8 | 805 | 74 | |
XM_008258416.2 | XP_008256638.1 | 8 - 9 | 926 | 74 | ||
XM_008258417.2 | XP_008256639.1 | 8 - 9 | 931 | 74 | ||
GenBank mRNA | U49933.1 | - | 6 - 7 | 787 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
