Assay Details
Gene Symbol: ST14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008259188.2 | XP_008257410.1 | 11 - 12 | 1510 | 81 | |
GenBank mRNA | EF472909.1 | - | 1 - 2 | 69 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
