Assay Details
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017347820.1 | XP_017203309.1 | 20 - 21 | 4256 | 110 | |
GenBank mRNA | EU407475.1 | - | 3 - 4 | 345 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
