Assay Details
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082034.1 | NP_001075503.1 | 3 - 4 | 392 | 70 | |
XM_008260198.2 | XP_008258420.1 | 4 - 5 | 602 | 70 | ||
GenBank mRNA | AF050174.1 | - | 3 - 4 | 392 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
