Assay Details
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082073.1 | NP_001075542.1 | 2 - 3 | 243 | 98 | |
GenBank mRNA | AF204927.1 | - | 2 - 3 | 243 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
