Assay Details
Gene Symbol: MGST3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082138.1 | NP_001075607.1 | 2 - 3 | 194 | 108 | |
XM_008263951.2 | XP_008262173.1 | 4 - 5 | 350 | 108 | ||
XM_008263952.2 | XP_008262174.1 | 3 - 4 | 305 | 108 | ||
GenBank mRNA | AY050567.1 | - | 2 - 3 | 194 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
