Assay Details
Gene Symbol: CD86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082208.1 | NP_001075677.1 | 4 - 5 | 715 | 79 | |
GenBank mRNA | D49842.1 | - | 4 - 5 | 715 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
