Assay Details
Gene Symbol: NEDD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082212.1 | NP_001075681.1 | 2 - 3 | 100 | 75 | |
GenBank mRNA | DQ067448.1 | - | 2 - 3 | 100 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
