Assay Details
Gene Symbol: NEDD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082212.1 | NP_001075681.1 | 3 - 4 | 177 | 65 | |
GenBank mRNA | DQ067448.1 | - | 3 - 4 | 177 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
