Assay Details
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082214.1 | NP_001075683.1 | 4 - 5 | 494 | 134 | |
GenBank mRNA | DQ120011.1 | - | 4 - 5 | 494 | 134 | |
GQ848313.1 | - | 4 - 5 | 494 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
