Assay Details
Gene Symbol: MYOZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082215.1 | NP_001075684.1 | 4 - 5 | 661 | 81 | |
XM_008269905.1 | XP_008268127.1 | 5 - 6 | 789 | 81 | ||
GenBank mRNA | DQ143044.1 | - | 4 - 5 | 661 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
