Assay Details
Gene Symbol: AKR1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082287.1 | NP_001075756.1 | 4 - 5 | 649 | 84 | |
GenBank mRNA | J05048.1 | - | 4 - 5 | 387 | 84 | |
M32818.1 | - | 4 - 5 | 649 | 84 | ||
U12316.1 | - | 4 - 5 | 465 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
