Assay Details
Gene Symbol: CXCL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082293.1 | NP_001075762.1 | 1 - 2 | 97 | 81 | |
GenBank mRNA | KT216053.1 | - | 1 - 2 | 68 | 81 | |
M57439.1 | - | 1 - 2 | 97 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
