Assay Details
Gene Symbol: TLR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082732.2 | NP_001076201.1 | 292 | 76 | ||
GenBank mRNA | AY101394.1 | - | 296 | 76 |

More Information
Set Membership: |
![]() ![]() |
