Assay Details
Gene Symbol: ELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082655.1 | NP_001076124.1 | 35 - 36 | 3939 | 78 | |
GenBank mRNA | AF388202.1 | - | 35 - 36 | 3939 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
