Assay Details
Gene Symbol: ETS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082661.1 | NP_001076130.1 | 4 - 5 | 598 | 102 | |
GenBank mRNA | AY561276.1 | - | 4 - 5 | 598 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
