Assay Details
Gene Symbol: ETS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082661.1 | NP_001076130.1 | 6 - 7 | 1107 | 84 | |
GenBank mRNA | AY561276.1 | - | 6 - 7 | 1107 | 84 | |
EU939724.1 | - | 2 - 3 | 344 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
