Assay Details
Gene Symbol: ETS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082661.1 | NP_001076130.1 | 7 - 8 | 1224 | 94 | |
GenBank mRNA | AY561276.1 | - | 7 - 8 | 1224 | 94 | |
EU939724.1 | - | 3 - 4 | 461 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
