Assay Details
Gene Symbol: PTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082680.1 | NP_001076149.1 | 544 | 84 | ||
GenBank mRNA | X73479.1 | - | 544 | 84 |

More Information
Set Membership: |
![]() ![]() |
