Assay Details
Gene Symbol: SLC25A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082686.1 | NP_001076155.1 | 1 - 2 | 124 | 61 | |
GenBank mRNA | AB009386.1 | - | 1 - 2 | 124 | 61 |
Gene Symbol: LOC100349768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715921.3 | XP_002715967.1 | 1 | 229 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
