Assay Details
Gene Symbol: PPM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082698.1 | NP_001076167.1 | 2 - 3 | 961 | 89 | |
XM_008271817.2 | XP_008270039.1 | 3 - 4 | 1024 | 89 | ||
XM_008271818.2 | XP_008270040.1 | 3 - 4 | 1021 | 89 | ||
XM_008271819.2 | XP_008270041.1 | 3 - 4 | 1063 | 89 | ||
XM_008271820.2 | XP_008270042.1 | 3 - 4 | 1063 | 89 | ||
XM_017349342.1 | XP_017204831.1 | 4 - 5 | 1186 | 89 | ||
GenBank mRNA | S87757.1 | - | 2 - 3 | 961 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
