Assay Details
Gene Symbol: LPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101689.1 | NP_001095159.1 | 5 - 6 | 1023 | 132 | |
GenBank mRNA | X07995.1 | - | 5 - 6 | 1023 | 132 |
Gene Symbol: LCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712428.3 | XP_002712474.1 | 5 - 6 | 981 | 132 | |
GenBank mRNA | Z27166.1 | - | 964 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() |
