Assay Details
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105673.1 | NP_001099143.1 | 2 - 3 | 518 | 108 | |
XM_008258588.2 | XP_008256810.1 | 3 - 4 | 683 | 108 | ||
XM_008258591.2 | XP_008256813.1 | 3 - 4 | 683 | 108 | ||
XM_017341452.1 | XP_017196941.1 | 3 - 4 | 683 | 108 | ||
XM_017341501.1 | XP_017196990.1 | 3 - 4 | 684 | 108 | ||
GenBank mRNA | EU014879.1 | - | 2 - 3 | 518 | 108 | |
JN541227.1 | - | 2 - 3 | 518 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
