Assay Details
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105673.1 | NP_001099143.1 | 3 - 4 | 744 | 75 | |
XM_008258588.2 | XP_008256810.1 | 4 - 5 | 909 | 75 | ||
GenBank mRNA | EU014879.1 | - | 3 - 4 | 744 | 75 | |
JN541226.1 | - | 3 - 4 | 744 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
