Assay Details
Gene Symbol: IL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122940.1 | NP_001116412.1 | 4 - 5 | 357 | 144 | |
XM_008258602.2 | XP_008256824.1 | 5 - 6 | 664 | 144 | ||
XM_008258609.2 | XP_008256831.1 | 4 - 5 | 349 | 144 | ||
GenBank mRNA | EU419127.1 | - | 4 - 5 | 357 | 144 | |
KT216067.1 | - | 4 - 5 | 357 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
