Assay Details
Gene Symbol: IL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163177.1 | NP_001156649.1 | 3 - 4 | 343 | 68 | |
NM_001171106.1 | NP_001164577.1 | 2 - 3 | 309 | 68 | ||
XM_008254869.2 | XP_008253091.1 | 3 - 4 | 358 | 68 | ||
GenBank mRNA | AF169169.1 | - | 3 - 4 | 399 | 68 | |
AF169170.1 | - | 2 - 3 | 309 | 68 | ||
DQ852343.1 | - | 3 - 4 | 343 | 68 | ||
EF606761.1 | - | 3 - 4 | 343 | 68 | ||
EU639687.1 | - | 3 - 4 | 343 | 68 | ||
KT216051.1 | - | 3 - 4 | 343 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
