Assay Details
Gene Symbol: GCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718258.3 | XP_002718304.1 | 4 - 5 | 689 | 78 | |
GenBank mRNA | FJ687583.1 | - | 4 - 5 | 370 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
