Assay Details
Gene Symbol: UGT2C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195805.1 | NP_001182734.1 | 5 - 6 | 1311 | 87 | |
GenBank mRNA | L01083.1 | - | 5 - 6 | 1230 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
