Assay Details
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163180.1 | NP_001156652.1 | 1 - 2 | 184 | 97 | |
GenBank mRNA | AF068057.1 | - | 1 - 2 | 184 | 97 | |
DQ852342.1 | - | 1 - 2 | 137 | 97 | ||
EF586071.1 | - | 1 - 2 | 137 | 97 | ||
EU639406.1 | - | 1 - 2 | 137 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
