Assay Details
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008265713.2 | XP_008263935.1 | 4 - 5 | 840 | 62 | |
GenBank mRNA | EU302911.1 | - | 1 - 2 | 61 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
