Assay Details
Gene Symbol: LOC100009480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082723.1 | NP_001076192.1 | 258 | 100 | ||
GenBank mRNA | AF308620.1 | - | 258 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
