Assay Details
Gene Symbol: NPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163113.1 | NP_001156585.1 | 86 | 92 | ||
XM_017350758.1 | XP_017206247.1 | 128 | 92 | |||
GenBank mRNA | EU978456.1 | - | 86 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
