Assay Details
Gene Symbol: UCP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171077.1 | NP_001164548.1 | 2 - 3 | 445 | 64 | |
GenBank mRNA | X14696.1 | - | 445 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
