Assay Details
Gene Symbol: LOC100328937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171121.1 | NP_001164592.1 | 981 | 63 | ||
XM_008252402.2 | XP_008250624.1 | 1057 | 63 | |||
XM_017339812.1 | XP_017195301.1 | 1058 | 63 | |||
GenBank mRNA | M36538.1 | - | 981 | 63 | ||
X05686.1 | - | 1000 | 63 | |||
X13853.1 | - | 982 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
