Assay Details
Gene Symbol: LOC100328937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171121.1 | NP_001164592.1 | 1196 | 66 | ||
XM_008252402.2 | XP_008250624.1 | 1272 | 66 | |||
GenBank mRNA | M11728.1 | - | 893 | 66 | ||
M36538.1 | - | 1196 | 66 | |||
X05686.1 | - | 1215 | 66 | |||
X13853.1 | - | 1197 | 66 |

More Information
Set Membership: |
![]() ![]() |
