Assay Details
Gene Symbol: TNFAIP8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171296.1 | NP_001164767.1 | 1 | 455 | 70 | |
XM_008264013.2 | XP_008262235.1 | 2 | 1304 | 70 | ||
XM_008264014.2 | XP_008262236.1 | 2 | 1250 | 70 | ||
XM_008264015.2 | XP_008262237.1 | 2 | 1280 | 70 | ||
XM_017345509.1 | XP_017200998.1 | 2 | 1229 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
