Assay Details
Gene Symbol: FLNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171371.1 | NP_001164842.1 | 5411 | 77 |
Gene Symbol: RA_M006_JSM7BED4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008250283.2 | XP_008248505.1 | 330 | 77 |

More Information
Set Membership: |
![]() ![]() |
