Assay Details
Gene Symbol: ELAVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185148.2 | NP_001172077.1 | 175 | 81 | ||
XM_008249127.2 | XP_008247349.1 | 454 | 81 | |||
GenBank mRNA | HM037267.1 | - | 176 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
