Assay Details
Gene Symbol: LOC100347603 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017349788.1 | XP_017205277.1 | 7 - 8 | 536 | 68 | |
XM_017349789.1 | XP_017205278.1 | 7 - 8 | 536 | 68 | ||
XM_017349790.1 | XP_017205279.1 | 7 - 8 | 536 | 68 | ||
XM_017349791.1 | XP_017205280.1 | 7 - 8 | 536 | 68 | ||
XM_017349792.1 | XP_017205281.1 | 7 - 8 | 536 | 68 | ||
XM_017349793.1 | XP_017205282.1 | 7 - 8 | 536 | 68 | ||
XM_017349794.1 | XP_017205283.1 | 7 - 8 | 536 | 68 | ||
XM_017349795.1 | XP_017205284.1 | 7 - 8 | 536 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
